3-Bromo-6-(trifluoromethyl)pyridazine 97%

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-6-(trifluoromethyl)-1,2-diazine

PC48565-250MG
APOSPC48565-250MGEA 166 EUR
APOSPC48565-250MG
3-Bromo-6-(trifluoromethyl)pyridazine 97%
3-Bromo-6-(trifluoromethyl)pyridazine
Formel: C₅H₂BrF₃N₂
Molecular Weight: 226,982 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14702751
CAS-Nummer: 174607-37-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR