5-[3-(Trifluoromethyl)phenyl]cyclohexane-1,3-dione

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(3,5-Dioxocyclohex-1-yl)benzotrifluoride

PC48581-1G
APOSPC48581-1GEA 466 EUR
APOSPC48581-1G
5-[3-(Trifluoromethyl)phenyl]cyclohexane-1,3-dione
5-[3-(Trifluoromethyl)phenyl]cyclohexane-1,3-dione
Formel: C₁₃H₁₁F₃O₂
Molecular Weight: 256,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00111873
CAS-Nummer: 144128-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR