3-(3-Methylbenzoyl)-1,1,1-trifluoroacetone 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(m-Tolyl)-4,4,4-trifluorobutane-1,3-dione, 4,4,4-Trifluoro-1-(m-tolyl)butane-1,3-dione, 1-(3-Methylphenyl)-4,4,4-trifluorobutane-1,3-dione

PC48584-250MG
APOSPC48584-250MGEA 68.3 EUR
APOSPC48584-250MG
3-(3-Methylbenzoyl)-1,1,1-trifluoroacetone 97%
3-(3-Methylbenzoyl)-1,1,1-trifluoroacetone
Formel: C₁₁H₉F₃O₂
Molecular Weight: 230,183 g/mol
Siedepunkt: 244 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11849497
CAS-Nummer: 53764-99-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR