3-(2-Methylbenzoyl)-1,1,1-trifluoroacetone 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,4,4-Trifluoro-1-(o-tolyl)butane-1,3-dione, 1-(o-Tolyl)-4,4,4-trifluorobutane-1,3-dione, 1-(2-Methylphenyl)-4,4,4-trifluorobutane-1,3-dione

PC48585-250MG
APOSPC48585-250MGEA 26.1 EUR
APOSPC48585-250MG
3-(2-Methylbenzoyl)-1,1,1-trifluoroacetone 97%
3-(2-Methylbenzoyl)-1,1,1-trifluoroacetone
Formel: C₁₁H₉F₃O₂
Molecular Weight: 230,183 g/mol
Siedepunkt: 97…103 °C (25 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD03420671
CAS-Nummer: 163266-02-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR