4-Nitrobenzenesulphonyl Fluoride 97+%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Nitrobenzene-1-sulphonyl fluoride

PC48588-5G
APOSPC48588-5GEA 60.9 EUR
APOSPC48588-5G
4-Nitrobenzenesulphonyl Fluoride 97+%
4-Nitrobenzenesulphonyl Fluoride
Formel: C₆H₄FNO₄S
Molecular Weight: 205,16 g/mol
Schmelzpunkt: 76…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009637
CAS-Nummer: 349-96-2
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR