1-(4-Fluorophenyl)cyclopropylamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Fluorophenyl)cyclopropanamine

PC48597-1G
APOSPC48597-1GEA 183 EUR
APOSPC48597-1G
1-(4-Fluorophenyl)cyclopropylamine
1-(4-Fluorophenyl)cyclopropylamine
Formel: C₉H₁₀FN
Molecular Weight: 151,181 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07374474
CAS-Nummer: 474709-83-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR