2-Chloro-4-iodo-6-(trifluoromethyl)pyridine 98%

Lieferant: Apollo Scientific

PC48602-1G
APOSPC48602-1GEA 21.9 EUR
APOSPC48602-1G
2-Chloro-4-iodo-6-(trifluoromethyl)pyridine 98%
2-Chloro-4-iodo-6-(trifluoromethyl)pyridine
Formel: C₆H₂ClF₃IN
Schmelzpunkt: 93…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD10699394
CAS-Nummer: 205444-22-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR