2,4-Difluoro-DL-phenylglycine hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-2-(2,4-difluorophenyl)acetic acid hydrochloride, Amino(2,4-difluorophenyl)acetic acid hydrochloride

PC48607-1G
APOSPC48607-1GEA 58 EUR
APOSPC48607-1G
2,4-Difluoro-DL-phenylglycine hydrochloride 95%
2,4-Difluoro-DL-phenylglycine hydrochloride
Formel: C₈H₇F₂NO₂·ClH
Molecular Weight: 223.604 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22380534
CAS-Nummer: 1374651-47-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR