3-Chlor-5-(trifluormethyl)phenylboronsäurepinakolester

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-5-(trifluoromethyl)benzeneboronic acid pinacol ester, 2-(3-Chloro-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PC48619-5G
APOSPC48619-5GEA 68.3 EUR
APOSPC48619-5G
3-Chlor-5-(trifluormethyl)phenylboronsäurepinakolester
3-Chlor-5-(trifluormethyl)phenylboronsäurepinakolester
Formel: C₁₃H₁₅BClF₃O₂
Molecular Weight: 306.52 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11856039
CAS-Nummer: 942069-65-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR