2,5-Difluorobenzeneboronic acid pinacol ester

Lieferant: Apollo Scientific

PC48624-1G
APOSPC48624-1GEA 119 EUR
APOSPC48624-1G
2,5-Difluorobenzeneboronic acid pinacol ester
2,5-Difluorobenzeneboronic acid pinacol ester
Formel: C₁₂H₁₅BF₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12405346
CAS-Nummer: 408492-25-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR