5-Brom-2-fluor-4-methylbenzonitril 98%

Lieferant: Apollo Scientific

PC48632-1G
APOSPC48632-1GEA 92.8 EUR
APOSPC48632-1G
5-Brom-2-fluor-4-methylbenzonitril 98%
5-Brom-2-fluor-4-methylbenzonitril
Formel: C₈H₅BrFN
Molecular Weight: 214,0300 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18452117
CAS-Nummer: 1269493-45-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR