tert-Butyl 3-(trifluoromethyl)-1,4,5,7-tetrahydro-6H-pyrazolo[3,4-c]pyridine-6-carboxylate

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 3-(trifluoromethyl)-4,5-dihydro-1H-pyrazolo[3,4-c]pyridine-6(7H)-carboxylate

PC48634-5G
APOSPC48634-5GEA 706 EUR
APOSPC48634-5G
tert-Butyl 3-(trifluoromethyl)-1,4,5,7-tetrahydro-6H-pyrazolo[3,4-c]pyridine-6-carboxylate
tert-Butyl 3-(trifluoromethyl)-1,4,5,7-tetrahydro-6H-pyrazolo[3,4-c]pyridine-6-carboxylate
Formel: C₁₂H₁₆F₃N₃O₂
Molecular Weight: 291,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044974
CAS-Nummer: 733757-89-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR