(6-Bromo-3-fluoropyridin-2-yl)methanol

Lieferant: Apollo Scientific
Achtung

PC48635-1G
APOSPC48635-1GEA 348 EUR
APOSPC48635-1G
(6-Bromo-3-fluoropyridin-2-yl)methanol
(6-Bromo-3-fluoropyridin-2-yl)methanol
Formel: C₆H₅BrFNO
Molecular Weight: 206,012 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16607006
CAS-Nummer: 918793-01-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR