2-(3-Hydroxy-n-propyl)-5-(trifluoromethyl)benzimidazole 97%
Lieferant: Apollo Scientific
PC4864-1G
APOSPC4864-1GEA
0
EUR
APOSPC4864-1G
2-(3-Hydroxy-n-propyl)-5-(trifluoromethyl)benzimidazole 97%
2-(3-Hydroxy-n-propyl)-5-(trifluoromethyl)benzimidazole
Formel:
C₁₁H₁₁F₃N₂O Schmelzpunkt: 139…141 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067735 CAS-Nummer: 175135-15-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...