(S)-2-Chloro-1-(4-fluorophenyl)ethanol 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC48643-1G
PC48643-250MG
APOSPC48643-1GEA
524
EUR
APOSPC48643-1G
APOSPC48643-250MG
(S)-2-Chloro-1-(4-fluorophenyl)ethanol 98%
(S)-2-Chloro-1-(4-fluorophenyl)ethanol
Formel:
C₈H₈ClFO Lagertemperatur: Raumtemperatur |
MDL:
MFCD07776653 CAS-Nummer: 126534-42-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...