1-(4-Chloro-3-fluorophenyl)ethan-1-ol
Lieferant: Apollo Scientific
|
Achtung
|
PC48645-1G
PC48645-250MG
APOSPC48645-1GEA
87
EUR
APOSPC48645-1G
APOSPC48645-250MG
1-(4-Chloro-3-fluorophenyl)ethan-1-ol
1-(4-Chloro-3-fluorophenyl)ethan-1-ol
Formel:
C₈H₈ClFO Lagertemperatur: Raumtemperatur |
MDL:
MFCD08461916 CAS-Nummer: 339001-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...