1-(4-Chloro-3-fluorophenyl)ethan-1-ol

Lieferant: Apollo Scientific
Achtung

PC48645-1G PC48645-250MG
APOSPC48645-1GEA 87 EUR
APOSPC48645-1G APOSPC48645-250MG
1-(4-Chloro-3-fluorophenyl)ethan-1-ol
1-(4-Chloro-3-fluorophenyl)ethan-1-ol
Formel: C₈H₈ClFO
Lagertemperatur: Raumtemperatur
MDL: MFCD08461916
CAS-Nummer: 339001-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR