6-Bromo-3-chloro-2,4-difluorobenzaldehyde 98%

Lieferant: Apollo Scientific

PC48651-1G
APOSPC48651-1GEA 377 EUR
APOSPC48651-1G
6-Bromo-3-chloro-2,4-difluorobenzaldehyde 98%
6-Bromo-3-chloro-2,4-difluorobenzaldehyde
Formel: C₇H₂BrClF₂O
Molecular Weight: 255.444 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11845975
CAS-Nummer: 1160573-22-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR