3-Bromo-4,5-difluorophenol 95%

Lieferant: Apollo Scientific
Achtung

PC48654-1G PC48654-250MG PC48654-5G
APOSPC48654-1GEA 522 EUR
APOSPC48654-1G APOSPC48654-250MG APOSPC48654-5G
3-Bromo-4,5-difluorophenol 95%
3-Bromo-4,5-difluorophenol
Formel: C₆H₃BrF₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD28556936
CAS-Nummer: 1807120-34-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR