6-(Trifluoromethyl)pyridine-2-carboxamidine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-(Trifluoromethyl)picolinimidamide hydrochloride
PC48657-1G
PC48657-250MG
APOSPC48657-1GEA
377
EUR
APOSPC48657-1G
APOSPC48657-250MG
6-(Trifluoromethyl)pyridine-2-carboxamidine hydrochloride
6-(Trifluoromethyl)pyridine-2-carboxamidine hydrochloride
Formel:
C₇H₆F₃N₃·ClH Molecular Weight: 225.599 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00662774 CAS-Nummer: 264884-49-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...