6-(Trifluoromethyl)pyridine-2-carboxamidine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-(Trifluoromethyl)picolinimidamide hydrochloride

PC48657-1G PC48657-250MG
APOSPC48657-1GEA 377 EUR
APOSPC48657-1G APOSPC48657-250MG
6-(Trifluoromethyl)pyridine-2-carboxamidine hydrochloride
6-(Trifluoromethyl)pyridine-2-carboxamidine hydrochloride
Formel: C₇H₆F₃N₃·ClH
Molecular Weight: 225.599 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00662774
CAS-Nummer: 264884-49-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR