7-Bromo-8-fluoroquinoline 95%

Lieferant: Apollo Scientific
Achtung

PC48659-1G
APOSPC48659-1GEA 699 EUR
APOSPC48659-1G
7-Bromo-8-fluoroquinoline 95%
7-Bromo-8-fluoroquinoline
Formel: C₉H₅BrFN
Molecular Weight: 226.045 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23163685
CAS-Nummer: 1375107-95-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR