4-Hydroxy-6-(trifluormethyl)-3-chinolincarbonsäure 97%

Lieferant: Apollo Scientific
Achtung

PC4869F-1G
APOSPC4869F-1GEA 23.2 EUR
APOSPC4869F-1G
4-Hydroxy-6-(trifluormethyl)-3-chinolincarbonsäure 97%
4-Hydroxy-6-(trifluoromethyl)quinoline-3-carboxylic acid
Formel: C₁₁H₆F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00236691
CAS-Nummer: 49713-47-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR