5-Iodo-4-methyl-2-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific
Achtung

PC48728-500MG
APOSPC48728-500MGEA 0 EUR
APOSPC48728-500MG
5-Iodo-4-methyl-2-(trifluoromethyl)pyridine
5-Iodo-4-methyl-2-(trifluoromethyl)pyridine
Formel: C₇H₅F₃IN
Lagertemperatur: Raumtemperatur
MDL: MFCD27934120
CAS-Nummer: 944317-55-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR