2-Bromo-5-fluoro-4-methylbenzaldehyde 98%

Lieferant: Apollo Scientific

PC48739-5G
APOSPC48739-5GEA 177 EUR
APOSPC48739-5G
2-Bromo-5-fluoro-4-methylbenzaldehyde 98%
2-Bromo-5-fluoro-4-methylbenzaldehyde
Formel: C₈H₆BrFO
Molecular Weight: 217,035 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07782047
CAS-Nummer: 916792-21-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR