5-Bromo-3-(trifluoromethyl)picolinic acid 95%

Lieferant: Apollo Scientific

PC48747-1G
APOSPC48747-1GEA 98.6 EUR
APOSPC48747-1G
5-Bromo-3-(trifluoromethyl)picolinic acid 95%
5-Bromo-3-(trifluoromethyl)picolinic acid
Formel: C₇H₃BrF₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD13185433
CAS-Nummer: 1211580-84-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR