4-Fluoro-1H-indole-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC4875-250MG
APOSPC4875-250MGEA
39.3
EUR
APOSPC4875-250MG
4-Fluoro-1H-indole-2-carboxylic acid
4-Fluoro-1H-indole-2-carboxylic acid
Formel:
C₉H₆FNO₂ Molecular Weight: 179,1500 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02664471 CAS-Nummer: 399-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...