5-Bromo-2-chloro-4-fluoronitrobenzene 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-4-chloro-2-fluoro-5-nitrobenzene
PC48769-5G
APOSPC48769-5GEA
49.3
EUR
APOSPC48769-5G
5-Bromo-2-chloro-4-fluoronitrobenzene 97%
5-Bromo-2-chloro-4-fluoronitrobenzene
Formel:
C₆H₂BrClFNO₂ Molecular Weight: 254.441 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12024826 CAS-Nummer: 1311197-88-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...