Perfluoro(1,3-dimethyl-5-ethylcyclohexane)
Lieferant: Apollo Scientific
|
Achtung
|
PC4877-100G
PC4877-25G
PC4877-5G
APOSPC4877-100GEA
458
EUR
APOSPC4877-100G
APOSPC4877-25G
APOSPC4877-5G
Perfluoro(1,3-dimethyl-5-ethylcyclohexane)
Perfluoro(1,3-dimethyl-5-ethylcyclohexane)
Formel:
C₁₀F₂₀ Siedepunkt: 144…146 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972337 CAS-Nummer: 155609-20-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...