2-Chloro-4-fluoro-5-iodonitrobenzene 96%

Lieferant: Apollo Scientific
Achtung

PC48773-1G PC48773-250MG PC48773-5G
APOSPC48773-1GEA 328 EUR
APOSPC48773-1G APOSPC48773-250MG APOSPC48773-5G
2-Chloro-4-fluoro-5-iodonitrobenzene 96%
2-Chloro-4-fluoro-5-iodonitrobenzene
Formel: C₆H₂ClFINO₂
Molecular Weight: 301.441 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30723635
CAS-Nummer: 2244086-60-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR