2,6-Difluorbenzylmercaptan

Lieferant: Apollo Scientific
Achtung

Synonyme: (2,6-Difluorophenyl)methanethiol

PC48785-250MG
APOSPC48785-250MGEA 65.4 EUR
APOSPC48785-250MG
2,6-Difluorbenzylmercaptan
2,6-Difluorbenzylmercaptan
Formel: C₇H₆F₂S
Lagertemperatur: Kühlschrank
MDL: MFCD06798019
CAS-Nummer: 878204-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR