3-Bromo-5-(trifluoromethoxy)phenylacetylene 98%

Lieferant: Apollo Scientific
Achtung

PC48787-1G PC48787-250MG PC48787-5G
APOSPC48787-1GEA 0 EUR
APOSPC48787-1G APOSPC48787-250MG APOSPC48787-5G
3-Bromo-5-(trifluoromethoxy)phenylacetylene 98%
3-Bromo-5-(trifluoromethoxy)phenylacetylene
Formel: C₉H₄BrF₃O
Lagertemperatur: Kühlschrank
MDL: MFCD31726319
CAS-Nummer: 2366994-52-5
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR