3-(Difluormethoxy)benzonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC4879-1G
PC4879-5G
PC4879-25G
APOSPC4879-1GEA
21.9
EUR
APOSPC4879-1G
APOSPC4879-5G
APOSPC4879-25G
3-(Difluormethoxy)benzonitril 98%
3-(Difluormethoxy)benzonitril
Formel:
C₈H₅F₂NO Molecular Weight: 169.1282 g/mol Siedepunkt: 224…225 °C Dichte: 1.26 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236225 CAS-Nummer: 97582-88-2 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...