4-Bromo-2-fluorophenyl acetate

Lieferant: Apollo Scientific
Achtung

PC48806-5G
APOSPC48806-5GEA 174 EUR
APOSPC48806-5G
4-Bromo-2-fluorophenyl acetate
4-Bromo-2-fluorophenyl acetate
Formel: C₈H₆BrFO₂
Molecular Weight: 233,034 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28156435
CAS-Nummer: 161424-78-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR