Ethyl 2-{2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]hydrazino}-2-oxoacetate 97%
Lieferant: Apollo Scientific
PC4881-1G
APOSPC4881-1GEA
640
EUR
APOSPC4881-1G
Ethyl 2-{2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]hydrazino}-2-oxoacetate 97%
Ethyl 2-{2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]hydrazino}-2-oxoacetate
Formel:
C₁₀H₉ClF₃N₃O₃ Molecular Weight: 311,645 g/mol Schmelzpunkt: 178…180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00215203 CAS-Nummer: 446276-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...