Ethyl 4,6-dioxo-5[3-(trifluoromethyl)phenyl]-1,3A,4,5,6,6A-hexahydropyrrolo[3,4-C]pyrazole-3-carboxylate

Lieferant: Apollo Scientific

PC4886-500MG
APOSPC4886-500MGEA 630 EUR
APOSPC4886-500MG
Ethyl 4,6-dioxo-5[3-(trifluoromethyl)phenyl]-1,3A,4,5,6,6A-hexahydropyrrolo[3,4-C]pyrazole-3-carboxylate
Ethyl 4,6-dioxo-5[3-(trifluoromethyl)phenyl]-1,3A,4,5,6,6A-hexahydropyrrolo[3,4-C]pyrazole-3-carboxylate
Formel: C₁₅H₁₂F₃N₃O₄
Schmelzpunkt: 172…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01315600
CAS-Nummer: 321522-08-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR