Ethyl-4[(3-fluoroanilino)carbonyl]-5-methylisoxazole-3-carboxylate

Lieferant: Apollo Scientific

PC4887-500MG
APOSPC4887-500MGEA 630 EUR
APOSPC4887-500MG
Ethyl-4[(3-fluoroanilino)carbonyl]-5-methylisoxazole-3-carboxylate
Ethyl-4[(3-fluoroanilino)carbonyl]-5-methylisoxazole-3-carboxylate
Formel: C₁₄H₁₃FN₂O₄
Molecular Weight: 292,27 g/mol
Schmelzpunkt: 122…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00664783
CAS-Nummer: 338792-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR