4-(Difluoromethyl)-2-(trifluoromethyl)benzaldehyde 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC48917-1G
PC48917-250MG
APOSPC48917-1GEA
623
EUR
APOSPC48917-1G
APOSPC48917-250MG
4-(Difluoromethyl)-2-(trifluoromethyl)benzaldehyde 95%
4-(Difluoromethyl)-2-(trifluoromethyl)benzaldehyde
Formel:
C₉H₅F₅O Lagertemperatur: Raumtemperatur |
MDL:
MFCD31759999 CAS-Nummer: 1453070-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...