Ethyl-2-{3-[(4-fluoroanilino)carbonyl]-2-pyridinyl}acetate

Lieferant: Apollo Scientific
Achtung

PC4893-250MG
APOSPC4893-250MGEA 263 EUR
APOSPC4893-250MG
Ethyl-2-{3-[(4-fluoroanilino)carbonyl]-2-pyridinyl}acetate
Ethyl-2-{3-[(4-fluoroanilino)carbonyl]-2-pyridinyl}acetate
Formel: C₁₆H₁₅FN₂O₃
Molecular Weight: 302,31 g/mol
Schmelzpunkt: 91…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00974991
CAS-Nummer: 338392-11-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR