5-Fluoroquinolin-8-amine

Lieferant: Apollo Scientific

PC48935-1G
APOSPC48935-1GEA 652 EUR
APOSPC48935-1G
5-Fluoroquinolin-8-amine
5-Fluoroquinolin-8-amine
Formel: C₉H₇FN₂
Molecular Weight: 162.164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09263638
CAS-Nummer: 161038-18-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR