Ethyl-2-[(2-fluorobenzoyl)amino]acetate 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC4897-5G
APOSPC4897-5GEA
0
EUR
APOSPC4897-5G
Ethyl-2-[(2-fluorobenzoyl)amino]acetate 97%
Ethyl-2-[(2-fluorobenzoyl)amino]acetate
Formel:
C₁₁H₁₂FNO₃ Schmelzpunkt: 40…42 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00793744 CAS-Nummer: 304657-05-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...