Ethyl 4-(2-fluorobenzoyl)isoxazole-3-carboxylate

Lieferant: Apollo Scientific

PC4898-100MG
APOSPC4898-100MGEA 271 EUR
APOSPC4898-100MG
Ethyl 4-(2-fluorobenzoyl)isoxazole-3-carboxylate
Ethyl 4-(2-fluorobenzoyl)isoxazole-3-carboxylate
Formel: C₁₃H₁₀FNO₄
Schmelzpunkt: 52…54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00664776
CAS-Nummer: 338761-76-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR