3-Bromo-4-fluorobenzene-1,2-diamine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-3,4-diamino-1-fluorobenzene
PC49003-1G
PC49003-5G
PC49003-250MG
APOSPC49003-1GEA
40.6
EUR
APOSPC49003-1G
APOSPC49003-5G
APOSPC49003-250MG
3-Bromo-4-fluorobenzene-1,2-diamine
3-Bromo-4-fluorobenzene-1,2-diamine
Formel:
C₆H₆BrFN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18071059 CAS-Nummer: 1257535-06-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...