3-Bromo-4-fluorobenzene-1,2-diamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-3,4-diamino-1-fluorobenzene

PC49003-1G PC49003-5G PC49003-250MG
APOSPC49003-1GEA 40.6 EUR
APOSPC49003-1G APOSPC49003-5G APOSPC49003-250MG
3-Bromo-4-fluorobenzene-1,2-diamine
3-Bromo-4-fluorobenzene-1,2-diamine
Formel: C₆H₆BrFN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18071059
CAS-Nummer: 1257535-06-0
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR