2-Bromo-3-fluoro-6-nitrophenol 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Bromo-4-fluoro-2-hydroxynitrobenzene, 2-Bromo-1-fluoro-3-hydroxy-4-nitrobenzene

PC49004-1G PC49004-5G PC49004-25G
APOSPC49004-1GEA 27.7 EUR
APOSPC49004-1G APOSPC49004-5G APOSPC49004-25G
2-Bromo-3-fluoro-6-nitrophenol 95%
2-Bromo-3-fluoro-6-nitrophenol
Formel: C₆H₃BrFNO₃
Molecular Weight: 235.995 g/mol
Schmelzpunkt: 65…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18071060
CAS-Nummer: 103979-08-4
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR