2-Bromo-3-fluoro-6-nitrophenol 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Bromo-4-fluoro-2-hydroxynitrobenzene, 2-Bromo-1-fluoro-3-hydroxy-4-nitrobenzene
PC49004-1G
PC49004-5G
PC49004-25G
APOSPC49004-1GEA
27.7
EUR
APOSPC49004-1G
APOSPC49004-5G
APOSPC49004-25G
2-Bromo-3-fluoro-6-nitrophenol 95%
2-Bromo-3-fluoro-6-nitrophenol
Formel:
C₆H₃BrFNO₃ Molecular Weight: 235.995 g/mol Schmelzpunkt: 65…66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD18071060 CAS-Nummer: 103979-08-4 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...