Ethyl 2-(hydrazinocarbonyl)-3-([3-(trifluoromethyl)anilino]carbonyl)cyclopropanecarboxylate

Lieferant: Apollo Scientific

PC4902-500MG
APOSPC4902-500MGEA 1180 EUR
APOSPC4902-500MG
Ethyl 2-(hydrazinocarbonyl)-3-([3-(trifluoromethyl)anilino]carbonyl)cyclopropanecarboxylate
Ethyl 2-(hydrazinocarbonyl)-3-([3-(trifluoromethyl)anilino]carbonyl)cyclopropanecarboxylate
Formel: C₁₅H₁₆F₃N₃O₄
Schmelzpunkt: 154…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01315623
CAS-Nummer: 318237-91-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR