3-Amino-7-(trifluoromethyl)-1H-indazole
Lieferant: Apollo Scientific
|
Achtung
|
PC49028-1G
APOSPC49028-1GEA
241
EUR
APOSPC49028-1G
3-Amino-7-(trifluoromethyl)-1H-indazole
3-Amino-7-(trifluoromethyl)-1H-indazole
Formel:
C₈H₆F₃N₃ Schmelzpunkt: 104…105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03426698 CAS-Nummer: 60330-35-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...