3-Amino-6-fluoroquinoline dihydrochloride

Lieferant: Apollo Scientific

Synonyme: 6-Fluoroquinolin-3-amine dihydrochloride

PC49030-1G
APOSPC49030-1GEA 607 EUR
APOSPC49030-1G
3-Amino-6-fluoroquinoline dihydrochloride
3-Amino-6-fluoroquinoline dihydrochloride
Formel: C₉H₇FN₂·2ClH
Molecular Weight: 235,086 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18375332
CAS-Nummer: 1266226-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR