3-Amino-8-fluoroquinoline dihydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Fluoroquinolin-3-amine dihydrochloride

PC49034-1G PC49034-250MG
APOSPC49034-1GEA 372 EUR
APOSPC49034-1G APOSPC49034-250MG
3-Amino-8-fluoroquinoline dihydrochloride
3-Amino-8-fluoroquinoline dihydrochloride
Formel: C₉H₇FN₂·2ClH
Molecular Weight: 235.086 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18375334
CAS-Nummer: 1266226-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR